Intermediate polar column test mix, certified reference material, Supelco®
Toimittaja: Merck
47301
47301.EA 58 EUR
47301.
Intermediate polar column test mix, certified reference material, Supelco®
Standardiaineet ja vertailumateriaalit Petroleum Industry Standards
Analytes:
Decane; 2,6-Dimethylaniline; 2,6-Dimethylphenol; Dodecane; 1-Octanol; 2-Octanone; Tetradecane; Tridecane; Undecane
Sertifikaatit: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.