VALEGRO Recirculating Coolers
Toimittaja: JULABO GmbH
9610050 9610035
473-3201EA 3970 EUR
473-3201 473-0040
VALEGRO Recirculating Coolers
Kiertotermostaatit Suljetut termostaattihaudejärjestelmät Jäähdyttävät termostaattihauteet
VALEGRO recirculating coolers are environmentally-friendly all-rounders for a wide range of temperature control tasks. They are characterised by a good ratio between cooling capacity and unit size as well as optimal usability. They work with natural refrigerant and can be adapted to application requirements thanks to their adjustable pump.
  • Climate-friendly thanks to natural refrigerant
  • OLED touch display with intuitive menu navigation
  • Good ratio between cooling capacity and size
  • USB-C and RS232 as standard, ethernet available as option
  • Illuminated, easy-to-