Hopeakloridi 4,9% in water, nanoparticle dispersion <200nm APS NW

Toimittaja: Thermo Fisher Scientific

47280.22 47280.36
47280.22EA 551 EUR
47280.22 47280.36
Hopeakloridi 4,9% in water, nanoparticle dispersion <200nm APS NW
Hopeakloridi

CAS No.: 7783-90-6 Molecular Formula: AgCl Formula Weight: 143.32 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.038 Appearance: White opaque Odour: Odourless Merck Reference: 14,8509 UN No.: 3082 MDL No.: MFCD00003399

Kaava: AgCl
Tiheys: 1,038
Storage Temperature: Jääkaappi
MDL numero: MFCD00003399
CAS-numero: 7783-90-6
EINECS: 232-033-3
UN: 3082
ADR: 9,III
Merck Index: 14,8509

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR