Mangaani(II)fluoridi ≥99.99% (metals basis), Puratronic®

Toimittaja: Thermo Fisher Scientific
Vaara

47109.14 47109.22
47109.14EA 565 EUR
47109.14 47109.22
Mangaani(II)fluoridi ≥99.99% (metals basis), Puratronic®
Mangaani(II)fluoridi

Purity: 99.99% CAS No.: 7782-64-1 Molecular Formula: F2Mn Formula Weight: 92.93 Physical Form: Crystalline Powder Density: 3.98 Sensitivity: Hygroscopic Appearance: Red Melting Point: 856° Solubility: Insoluble in water, alcohol. Soluble in dilute HF, conc. HCl or HNO3 UN No.: UN3288 MDL No.: MFCD00016222

Kaava: F₂Mn
Sulamispiste: 856 °C
Tiheys: 3,98
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016222
CAS-numero: 7782-64-1
EINECS: 231-960-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,5729

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR