Performance and Perfection laboratory freezers with plastic inner liner, SFFsg and SFFvh
Toimittaja: Liebherr
SFFVH5501 SFFSG4001_740 SFFSG5501_740 SFFVH4001
LIEBSFFVH5501EA 0 EUR
LIEBSFFVH5501 LIEBSFFSG4001_740 LIEBSFFSG5501_740 471-1341
Performance and Perfection laboratory freezers with plastic inner liner, SFFsg and SFFvh
Pakastimet Laboratory / General Purpose Freezers
Performance series with no lighting and perfection series with the LED front lighting provides optimal illumination of the drawers even in dark environments, for the perfect overview, the desired substance is immediately at hand, the door can be quickly closed again, and the interior temperature remains stable. For low consumption and lower energy costs.
  • Alarm relay, alarm test function and custom alarm settings possible
  • Self-closing door at <90° and mechanical lock provided
  • Antimicrobial handle with opening mechanism
  • Performance series with super-