Table top combined refrigerators/freezers
Toimittaja: Liebherr
KTS102 KTS126 KTS148 KTS165
471-0993EA 0 EUR
471-0993 471-0994 471-0995 LIEBKTS165
Table top combined refrigerators/freezers
Jääkaapit
Table top refrigerators for general use.
  • Static cooling
  • Automatic defrost
  • 'EasyClean' interior provides full modularity and ultra-smooth walls for easy cleaning
  • Adjustable feet
  • Reversible door
  • Glass shelves
Temperature range: −3 to +8 °C