1-Chloroethylisopropyl carbonate 98%

Toimittaja: Thermo Fisher Scientific
Vaara

470930250 470931000
470930250.EA 36.3 EUR
470930250. 470931000.
1-Chloroethylisopropyl carbonate 98%
1-Chloroethylisopropyl carbonate
Kaava: C₆H₁₁ClO₃
Kiehumispiste: 164,3°C
Tiheys: 1,09
Leimahduspiste: 55°C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD04038100
CAS-numero: 98298-66-9
UN: 2920
ADR: 8,II

Order Now

Testitulosten erittely

Appearance colorless to almost colorless liquid
Infrared spectrum Conforms
GC ≥97.5 %
Refractive index 1.4110 to 1.4150 (20°C, 589 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR