Natriumtiosulfaatti concentrate for dilution to 0.1 N vesiliuos, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Toimittaja: Avantor
4695.EA 26.48 EUR
4695.
Natriumtiosulfaatti concentrate for dilution to 0.1 N vesiliuos, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Natriumtiosulfaatti
Kaava: Na₂S₂O₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003499
CAS-numero: 7772-98-7
EINECS: 231-867-5
Order Now
Testitulosten erittely
For Laboratory,Research,or Manufacturing Use 0.0998-0.1002
Normality (by titrimetry) Passes Test
(Makes 0.1N solution after dilution to 1000 mL)