CTK1D7390 97%, Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY

Synonyms: DiI, DiI perchlorate

M-Clarity™ quality level = MQ100

468495-100MG
468495-100MGEA 363 EUR
468495-100MG
CTK1D7390 97%, Sigma-Aldrich®
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate

1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate is also called as Dil. It can be used to study HCV (Hepatitis C virus) cell interactions in the HCV entry process. It is used to label oxidized LDL (low-density lipoprotein) to quantify the uptake of DiI-oxLDL in human endothelial cells.

Kaava: C₅₉H₉₇ClN₂O₄
Sulamispiste: 68 °C (haj. lämpötila, lit.)
Storage Temperature: Jääkaappi
MDL numero: MFCD00142354
CAS-numero: 41085-99-8

Order Now

Testitulosten erittely

Appearance (Colour) Red to Very Dark Red and Purple to Very Dark Purple
Appearance (Form) Powder to Crystalline Powder
Infrared spectrum Conforms to Structure
Wavelength 276 - 282 nm
Extinction Coefficient ≥70000
Extinction Coefficient, c = 0.01 g/L; Methanol ≥10000
Carbon 73.2 - 78.5%
Nitrogen 2.7 - 3.3%
Purity (HPLC) ≥96.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR