Polyoxyethylene bis(amine), M.W. 1,000

Toimittaja: Thermo Fisher Scientific

46833.MD 46833.03 46833.06
46833.MDEA 121 EUR
46833.MD 46833.03 46833.06
Polyoxyethylene bis(amine), M.W. 1,000
Polyoxyethylene bis(amine)

CAS No.: 24991-53-5 Molecular Formula: H2NCH2CH2(OCH2CH2)nNH2 Physical Form: Granular solid Sensitivity: Air Sensitive Appearance: White to off-white MDL No.: MFCD00132004

Kaava: H₂NCH₂CH₂(OCH₂CH₂)nNH₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00132004
CAS-numero: 24991-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR