Tetrabutylammonium dihydrogen phosphate ≥98%, non-UV

Toimittaja: Thermo Fisher Scientific
Varoitus

46746.04 46746.09 46746.18
46746.04EA 195 EUR
46746.04 46746.09 46746.18
Tetrabutylammonium dihydrogen phosphate ≥98%, non-UV
Tetrabutylammonium dihydrogen phosphate

Tetra-n-butylammonium phosphate, is used in chemicals, pharmaceutical research, pharmaceutical intermediates, and reagents. Tetrabutylammonium phosphate can be used in organic synthesis. It is used as an Ion-associating reagent for HPLC.

Kaava: C₁₆H₃₈NO₄P
Sulamispiste: 151…154 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064526
CAS-numero: 5574-97-0
EINECS: 226-947-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR