4'-Trifluoromethylbiphenyl-4-carbaldehyde

Toimittaja: Thermo Fisher Scientific
Varoitus

467130010 467130050
467130010.EA 131 EUR
467130010. 467130050.
4'-Trifluoromethylbiphenyl-4-carbaldehyde
4'-Trifluoromethylbiphenyl-4-carbaldehyde
Kaava: C₁₄H₉F₃O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01862519
CAS-numero: 90035-34-0

Order Now

Testitulosten erittely

Appearance white to light yellow powder or crystals
Infrared spectrum Conforms
GC ≥97.5 %
Melting point 74°C to 78°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR