Desiccator cabinets
Toimittaja: SciLabware
DWA301
467-0004EA 439 EUR
467-0004
Desiccator cabinets
Eksikaattorit ja kaappieksikaattorit Kaappieksikaattori
Clear, high impact polystyrene cabinets have an airtight door with foam rubber gasket.
  • Cabinets can be stacked to save space
Toimitustiedot: Supplied with sealed jar of silica gel.