Drying ovens with forced convection FED series, Avantgarde.Line
Toimittaja: Binder
9010-0295 9010-0293 9010-0299 9010-0301
466-0482EA 2800 EUR
466-0482 466-0483 466-0484 BIND9010-0301
Drying ovens with forced convection FED series, Avantgarde.Line
Lämpökaapit Uunit, puhallinavusteinen ja luonnollinen ilmankierto
With forced convection and enhanced timer functions, the Avantgarde.Line heating and drying chambers offer quality and process reliability. With a wide temperature range up to 300 °C, these drying ovens can be used in all applications. Short heat up times and large power reserves are ideal for applications in science, research and industry. The APT.line™ preheating chamber technology gives homegeneous temperature distribution and excellent temperature uniformity and fluctuation. Units up to 115 litres can be stacked on top of each other to save space. The units have 60 mm insulation whichMore Product Information