Sudan red 7B (fat red bluish, solvent red 19, fat red 7B), Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY

Synonyms: N-Ethyl-1-((4-(phenyldiazenyl)phenyl)diazenyl)naphthalen-2-amine

46290-10G 46290-50G
46290-10GEA 107 EUR
46290-10G 46290-50G
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B), Sigma-Aldrich®
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Kaava: C₂₄H₂₁N₅
MW: 379.4570 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003904
CAS-numero: 6368-72-5
EINECS: 228-862-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e8bcad8994d0be85b60671e3b670c01fcb343e7&vid=4e8bcad8994d0be85b60671e3b670c01fcb343e7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR