Circulators, NE4 series, Clifton
Toimittaja: NICKEL ELECTRO
NE4-8P NE4-14P NE4-22P NE4-28P NE4-14HT NE4-22HT NE4-8HT NE4-56P
462-0049EA 2420 EUR
462-0049 462-0044 462-0039 462-0047 462-0113 462-0114 462-0112 NICKNE4-56P
Circulators, NE4 series, Clifton
Kiertotermostaatit Avoimet termostaattihaudejärjestelmät Hauteet, joissa irrotettavat termostaattihauteet
These circulating water baths are available in two ranges for circulating just water up to 99 °C or oil up to 130 °C. Operating temperatures below ambient can be achieved using an optional refrigerated dip cooler. Units have touch pad controls with easy wipe clean fronts which are chemical resistant and splash proof.
  • Powerful circulation pump, provides a constant flow
  • Convenient ‘push-fit and lock' fittings on both inlet and outlet ports for secure external water circulation
  • Control options, single point temperature control or process control with two
More Product Information
New Product