Heating circulators, DYNEO™ DD
Toimittaja: JULABO GmbH
9021504 9021506 9021512 9021526 9021506.A 9021512.A 9021526.A 9021504.A 9021512.D 9021526.D 9021504.D 9021506.D
461-1384EA 3032 EUR
461-1384 461-1385 461-1386 461-1387 461-1398 461-1399 461-1400 461-1397 461-1413 461-1414 461-1410 461-1412
Heating circulators, DYNEO™ DD
Kiertotermostaatit Avoimet termostaattihaudejärjestelmät Hauteet, joissa sisäänrakennetut termostaattihauteet
The DYNEO™ DD heating circulators are available in several bath combinations, with varying bath sizes. DYNEO is designed for internal as well as external applications. Operation and handling is easy using the large TFT colour display and the state of the art turn and push controller. The bath tanks are well insulated with a built-in coil for counter cooling. An integrated drain tap makes emptying the tank safe and clean.
  • Easy to switch between internal and external circulation
  • Integrated M16x1 pump connectors M16x1
  • Multi-lingual user interface
  • Powerful and