Bath tanks, stainless steel, for CORIO C / CD
Toimittaja: JULABO GmbH
9903405 9903413 9903417 9903419 9903427 9903433 9903439
461-0921EA 302 EUR
461-0921 461-0922 461-0923 461-0924 461-0925 461-0926 461-0927
Bath tanks, stainless steel, for CORIO C / CD
Hauteet Vesihauteet, sekoittamattomat
Stainless steel bath tanks in different sizes. For use with all CORIO C and CD circulators.
Temperature range: +20...+150 °C