Recirculating cooler, AWC100
Toimittaja: JULABO GmbH
9630100
461-0324EA 1500 EUR
461-0324
Recirculating cooler, AWC100
Kiertotermostaatit Suljetut termostaattihaudejärjestelmät Jäähdyttävät termostaattihauteet
The compact AWC100 air to water recirculating cooler is designed to remove small heat loads from external systems where cooling water can be above ambient and where temperature control is secondary. Coolers which do not have a refrigeration unit are ideal for use in circuits with Peltier elements, analytical devices, polarimeters, refractometers and CCD cameras. The ventilator speed (i.e. the cooling capacity) can be adjusted in two stages, cooling capacity depends on the temperature difference between return line temperature and ambient temperature. The cooling principle: Warm bath fluid isMore Product Information