Digital block heaters
Toimittaja: Thermo Fisher Scientific
88870004 88870005 88870006
460-0306EA 490 EUR
460-0306 460-0307 460-0308
Digital block heaters
Kuivahauteet Digitaaliset kuivahauteet
Versatile digital block heaters offer a wide range of blocks to accommodate various tubes provide temperature uniformity and stability for reproducible results. The units have digital controls and display of time and temperature and are durable with a powder-coated steel body. Ideal for sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation. Choice of models to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications.
  • Built-in temperature sensing probe for