Ytterbium ≥99.9% (REO, rare earth oxide basis), jauhe < 200 mesh

Toimittaja: Thermo Fisher Scientific
Vaara


Packaged under argon.

45920.04 45920.09 45920.18
45920.04EA 390 EUR
45920.04 45920.09 45920.18
Ytterbium ≥99.9% (REO, rare earth oxide basis), jauhe < 200 mesh
Ytterbium

Purity: 99.9% CAS No.: 7440-64-4 Physical Form: Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Silver Odour: Odourless UN No.: 3089 MDL No.: MFCD00011286

Kaava: Yb
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011286
CAS-numero: 7440-64-4
EINECS: 231-173-2
UN: 3089
ADR: 4.1,II

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR