Silikoniöljy high temperature , colourless liquid for oil baths

Toimittaja: Thermo Fisher Scientific

45896.A4 45896.22 45896.36
45896.A4EA 2210 EUR
45896.A4 45896.22 45896.36
Silikoniöljy high temperature , colourless liquid for oil baths
Silikoniöljy

Usable temperature range: 25 to 250 °C (open system) and 25 to 315 °C (closed system)


Refractive Index: 1.505


Applications: High temperature open system, closed system, circulating, closed loop, and heat transfer baths. High temperature fluids for laboratory research apparatus and instruments.

Kaava: [Si(CH3)2O]x[Si(C6H5)2O]k
Tiheys: 1,07 g/mL
Leimahduspiste: >300 °C(572 °F)
MDL numero: MFCD00132673
CAS-numero: 68083-14-7
Merck Index: 14,8495

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR