Optical windows
Toimittaja: Thermo Fisher Scientific
45387.KS 45386.KS 45560.KS 45666.KS 45813.KS 45856.KS 45925.KS 45986.KS 46057.KS 46089.KS
45387.KSEA 655 EUR
45387.KS 45386.KS 45560.KS 45666.KS 45813.KS 45856.KS 45925.KS 45986.KS 46057.KS 46089.KS
Optical windows
Optiset ikkunat
These optical windows are ideally stored at ambient temperatures.
  • Clear aperture 85%
  • Surface finish 60/40 or 40/20