Pesticide reference standard, Promecarb, PESTANAL®, analytical standard, Supelco®
Toimittaja: Merck
45634-50MG
45634-50MGEA 77.4 EUR
45634-50MG
Pesticide reference standard, Promecarb, PESTANAL®, analytical standard, Supelco®
Standardiaineet ja vertailumateriaalit Organic Standards
Promecarb may be used as a reference standard for the determination of promecarb in water by solid-phase extraction and liquid chromatography coupled with ionspray mass spectrometry.
  • Contains N-methylcarbamate group
  • Mode of action involves inhibiting the cholinesterase enzyme activity
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9365ee54f07b058d216b31d7fed02bd402222784&vid=9365ee54f07b058d216b31d7fed02bd402222784&prid=web&clid=SAPDAM