Bis(1,5-cyclooctadiene)rhodium(I) hexafluoroantimonate

Toimittaja: Thermo Fisher Scientific
Varoitus

45523.MD 45523.03
45523.MDEA 323 EUR
45523.MD 45523.03
Bis(1,5-cyclooctadiene)rhodium(I) hexafluoroantimonate
Bis(1,5-cyclooctadiene)rhodium(I) hexafluoroantimonate

CAS No.: 130296-28-5 Molecular Formula: C16H24F6RhSb Formula Weight: 555.01 Physical Form: Crystalline powder Appearance: Brown UN No.: 3077 MDL No.: MFCD11113031

Kaava: C₁₆H₂₄F₆RhSb
Storage Temperature: Huoneenlämpö
MDL numero: MFCD11113031
CAS-numero: 130296-28-5
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR