Muffle furnaces, Thermolyne™
Toimittaja: Thermo Fisher Scientific
F30420C-33 F30420C-33 F30420C-33-80 F30430CM-33 F48020-33 F6020C-33
BARNF30420C-33EA 0 EUR
BARNF30420C-33 455-0339 455-0343 455-0347 455-0341 455-0342
Muffle furnaces, Thermolyne™
Lämpökaapit Uuni
Thermolyne™ muffle furnaces offer heating solutions for a number of application needs, from general laboratory use to industrial applications. Units are ideal for fast heat up and temperature control up to 1200 °C. The furnaces are used for chemical analysis, materials testing, determination of volatiles in waste water treatment plants, industrial plants, pharmaceutical and cosmetic companies, environmental laboratories, agriculture laboratories and paper manufacturers.
  • Fast heat up times and outstanding energy efficiency
  • Range of capacity from 1,3 to 45