Selenium standard dissolved in oil, 1.000 µg/g Se in mineral oil Specpure® for wear metal analysis
Toimittaja: Thermo Fisher Scientific
45305.18
45305.18EA 199 EUR
45305.18
Selenium standard dissolved in oil, 1.000 µg/g Se in mineral oil Specpure® for wear metal analysis
Standardiaineet ja vertailumateriaalit Cationic Standards Selenium Standards
Physical form: liquid
MDL No.: MFCD00134090