Selenium 82, 82 Se, plasma standard solution, 82 Se 10 µg/ml, Specpure®

Toimittaja: Thermo Fisher Scientific

45284.ADEA 610 EUR
45284.AD
Selenium 82, 82 Se, plasma standard solution, 82 Se 10 µg/ml, Specpure®
Standardiaineet ja vertailumateriaalit Cationic Standards Selenium Standards

Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00084581

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR