Cadmium 106, 106 Cd, plasma standard solution, 106 Cd 10 µg/ml, Specpure®

Toimittaja: Thermo Fisher Scientific

45274.ADEA 774 EUR
45274.AD
Cadmium 106, 106 Cd, plasma standard solution, 106 Cd 10 µg/ml, Specpure®
Standardiaineet ja vertailumateriaalit Cationic Standards Cadmium Standards

Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00143885

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR