Platina 70% (nominal) on high surface area advanced carbon support, HiSPEC® 13100

Toimittaja: Thermo Fisher Scientific
Vaara

45041.06 45041.03
45041.06EA 0 EUR
45041.06 45041.03
Platina 70% (nominal) on high surface area advanced carbon support, HiSPEC® 13100
Platina

CAS No.: 7440-06-4 Appearance: Dark grey to black Odour: Odourless UN No.: UN1325 MDL No.: MFCD00011179

Kaava: Pt
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011179
CAS-numero: 7440-06-4
UN: 1325
ADR: 4.1,II

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR