Holmiumtrifluoridi, vedetön ≥99.9% (REO, rare earth oxide basis)

Toimittaja: Thermo Fisher Scientific
Vaara

45033.18 45033.09
45033.18EA 1220 EUR
45033.18 45033.09
Holmiumtrifluoridi, vedetön ≥99.9% (REO, rare earth oxide basis)
Holmiumtrifluoridi

Purity: 99.9% CAS No.: 13760-78-6 Molecular Formula: HoF3 Formula Weight: 221.93 Physical Form: Powder Density: 7.6 Melting Point: 1143° UN No.: 3288 MDL No.: MFCD00016142

Kaava: HoF₃
Sulamispiste: 1143 °C
Tiheys: 7,6
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016142
CAS-numero: 13760-78-6
EINECS: 237-352-1
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR