Rikkihappo 95% 95, ARISTAR® Trace_analysis

Toimittaja: VWR Chemicals
Vaara

450061QEA 153 EUR
450061Q
Rikkihappo 95% 95, ARISTAR® Trace_analysis
Rikkihappo
Kaava: H₂SO₄
MW: 98.08 g/mol
Kiehumispiste: 330 °C (1013 hPa)
Sulamispiste: 10.38 °C
Tiheys: 1.84 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064589
CAS-numero: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
REACH: 01-2119458838-20

Order Now

Testitulosten erittely

Assay Min. 95.0 %
Cl (Chloride) Max. 0.1 ppm
NO3 (Nitrate) Max. 0.1 ppm
PO4 (Phosphate) Max. 0.05 ppm
Al (Aluminium) Max. 0.050 ppm
As (Arsenic) Max. 0.005 ppm
B (Boron) Max. 0.050 ppm
Ba (Barium) Max. 0.050 ppm
Ca (Calcium) Max. 0.100 ppm
Cd (Cadmium) Max. 0.001 ppm
CO Max. 0.005 ppm
Cu (Copper) Max. 0.010 ppm
Fe (Iron) Max. 0.050 ppm
Hg (Mercury) Max. 0.001 ppm
K (Potassium) Max. 0.050 ppm
Mg (Magnesium) Max. 0.030 ppm
Mn (Manganese) Max. 0.005 ppm
Na (Sodium) Max. 0.300 ppm
NH4 (Ammonium) Max. 1.0 ppm
Ni (Nickel) Max. 0.005 ppm
Pb (Lead) Max. 0.005 ppm
Se (Selenium) Max. 0.100 ppm
Sr (Strontium) Max. 0.005 ppm
Zn (Zinc) Max. 0.020 ppm
Oxygen absorbing components Max. 1 ppm
Non-volatile matter Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR