Rodium 5% on activated carbon, paste C101023-5

Toimittaja: Thermo Fisher Scientific

44863.04 44863.09
44863.04EA 627 EUR
44863.04 44863.09
Rodium 5% on activated carbon, paste C101023-5
Rodium

CAS No.: 7440-16-6 Physical Form: Paste Appearance: Black MDL No.: MFCD03458392

Kaava: Rh
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011201
CAS-numero: 7440-16-6
EINECS: 231-125-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR