Silikageeli amorfinen ≥99.9% (metals basis), nanopowder

Toimittaja: Thermo Fisher Scientific
Varoitus

44781.14 44781.22 44781.36
44781.14EA 176 EUR
44781.14 44781.22 44781.36
Silikageeli amorfinen ≥99.9% (metals basis), nanopowder
Silikageeli amorfinen

Purity: 99.9% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: 80nm APS Powder Density: 2.4 Appearance: White Odour: Odourless pH: 6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Kaava: SiO₂
Kiehumispiste: 2230 °C
Sulamispiste: 1710 °C
Tiheys: 2,4
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011232
CAS-numero: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,8493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR