Poly(3-octylthiophene-2,5-diyl) regiorandom low metals

Toimittaja: Thermo Fisher Scientific

44778.01 44778.03
44778.01EA 449 EUR
44778.01 44778.03
Poly(3-octylthiophene-2,5-diyl) regiorandom low metals
Poly(3-octylthiophene-2,5-diyl)

CAS No.: 104934-51-2 Physical Form: Powder Solubility: Soluble in THF, CHCl3 MDL No.: MFCD00241412


Applications: Conducting polymers

Storage Temperature: Huoneenlämpö
MDL numero: MFCD00241412
CAS-numero: 104934-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR