Zirconium(IV) Propoxide ≥70% (w/w) in n-propanol

Toimittaja: Thermo Fisher Scientific
Vaara


Packaged under argon in resealable ChemSeal™ bottles.

44770.18 44770.30
44770.18EA 84.7 EUR
44770.18 44770.30
Zirconium(IV) Propoxide ≥70% (w/w) in n-propanol
Zirconium(IV) Propoxide

Packaged under Argon in resealable ChemSeal™ bottles


CAS No.: 23519-77-9 Molecular Formula: Zr[O(CH2)2CH3]4 Formula Weight: 327.56 Flash Point: 28°(83°F) Physical Form: Liquid Density: 1.044 Refractive Index: 1.4510 Sensitivity: Moisture Sensitive UN No.: UN1993 MDL No.: MFCD00015307

Kaava: C₁₂H₂₈O₄Zr
Tiheys: 1,044
Leimahduspiste: 28 °C (83 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00015307
CAS-numero: 23519-77-9
EINECS: 245-711-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR