Melting point standard, phenyl salicylate

Toimittaja: Merck

44770-5G
44770-5GEA 52.3 EUR
44770-5G
Melting point standard, phenyl salicylate
Standardiaineet ja vertailumateriaalit Sulamispistestandardit

Mettler-Toledo calibration substance ME 30034252, traceable to primary standards.


  • Boiling point: 172 to 173 °C/12 mm Hg (lit.)
  • Format: neat
  • Limited shelf life


Synonym: Phenyl salicylate, salol

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb097012497ce615dfd5ce235f15634686a11f19&vid=bb097012497ce615dfd5ce235f15634686a11f19&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR