Fast blue BB salt mikroskopiaan, histologiaan, Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY
Varoitus

44670-10G 44670-50G
44670-10GEA 30.8 EUR
44670-10G 44670-50G
Fast blue BB salt mikroskopiaan, histologiaan, Sigma-Aldrich®
Fast blue BB salt

Synonyms: 4-Amino-2,5-diethoxybenzanilide diazotated zinc double salt, 4-Benzoylamino-2,5-diethoxybenzenediazonium chloride hemi(zinc chloride) salt

Kaava: C₃₄H₃₆Cl₄N₆O₆Zn
MDL numero: MFCD00074765
CAS-numero: 5486-84-0
EINECS: 226-817-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a5533f11234363beaeaf1bc356010e195ee2bd&vid=11a5533f11234363beaeaf1bc356010e195ee2bd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR