Cesium trifluoroacetate

Toimittaja: Thermo Fisher Scientific
Varoitus

44633.09 44633.18
44633.09EA 109 EUR
44633.09 44633.18
Cesium trifluoroacetate
Cesium trifluoroacetate

CAS No.: 21907-50-6 Molecular Formula: CsOOCCF3 Formula Weight: 245.92 Physical Form: Crystalline Melting Point: 115° MDL No.: MFCD00051304

Kaava: C₂CsF₃O₂
Sulamispiste: 115 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00051304
CAS-numero: 21907-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR