Copper(II) trifluoroacetate hydrate

Toimittaja: Thermo Fisher Scientific
Varoitus

44624.09 44624.18
44624.09EA 223 EUR
44624.09 44624.18
Copper(II) trifluoroacetate hydrate
Copper(II) trifluoroacetate hydrate

CAS No.: 123333-88-0 Molecular Formula: Cu(OOCCF3)2.xH2O Formula Weight: 289.57(anhy) Physical Form: Crystalline MDL No.: MFCD00013200

Kaava: C₄CuF₆O₄·xH₂O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013200
CAS-numero: 123333-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR