Dragendorff'n reagenssi derivatisation reagent, TLC-analyyseihin, Supelco®

Toimittaja: Merck


M-Clarity™ quality level = MQ100

44578-100ML-F
44578-100ML-FEA 145 EUR
44578-100ML-F
Dragendorff'n reagenssi derivatisation reagent, TLC-analyyseihin, Supelco®
Dragendorff'n reagenssi

This non-aqueous Thiess, Reuther and Vagujfalvi variant of the reagent is used to detect alkaloids and other nitrogen compounds in TLC.


Composition: Acetic acid, ethyl acetate, sodium iodide, sodium tetraiodobismutate

Kiehumispiste: 77 °C (1013 hPa)
Tiheys: 1,2 g/cm³ (20 °C)
Leimahduspiste: –4 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00283106
CAS-numero: 39775-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR