Europium ≥99.9% (metals basis), sauvoja, Ø 9.52 mm (0.375 in)

Toimittaja: Thermo Fisher Scientific
Vaara

44545.HF 44545.DJ
44545.HFEA 990 EUR
44545.HF 44545.DJ
Europium ≥99.9% (metals basis), sauvoja, Ø 9.52 mm (0.375 in)
Europium

Purity: 99.9% CAS No.: 7440-53-1 Molecular Formula: Packaged in mineral oil Physical Form: ≈0.37g/mm, Cast rod, diameter approximate Appearance: Silver/gray Odour: Odourless UN No.: 3208 MDL No.: MFCD00010992

Kaava: Eu
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00010992
CAS-numero: 7440-53-1
EINECS: 231-161-7
UN: 3208
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR