Retinoic acid ≥98%

Toimittaja: Thermo Fisher Scientific
Varoitus

44540.02 44540.04 44540.77
44540.02EA 347 EUR
44540.02 44540.04 44540.77
Retinoic acid ≥98%
Retinoic acid

Purity: 98% CAS No.: 302-79-4 Molecular Formula: C20H28O2 Formula Weight: 300.44 Physical Form: Crystalline Sensitivity: Light Sensitive Beilstein Reference: 2057223 Merck Reference: 14,8165 MDL No.: MFCD00001551

Kaava: C₂₀H₂₈O₂
Sulamispiste: 180…182 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001551
CAS-numero: 302-79-4
EINECS: 206-129-0
Merck Index: 14,8165

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR