Sinkkioksidi ≥99% (metals basis), nanopowder

Toimittaja: Thermo Fisher Scientific
Varoitus

44533.09 44533.18
44533.09EA 295 EUR
44533.09 44533.18
Sinkkioksidi ≥99% (metals basis), nanopowder
Sinkkioksidi

Purity: 99% CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: 20-30nm APS Powder Density: 5.606 Appearance: White Odour: Odourless Melting Point: 1975° Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300

Kaava: ZnO
Sulamispiste: 1975 °C
Tiheys: 5,606
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011300
CAS-numero: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 14,10147

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR