Tebbe Reagent 0.5 M in toluene

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: µ-Chloro-µ-methylenebis(cyclopentadienyl)titaniumdimethylaluminum, Bis(cyclopentadienyl-mu-chloro(dimethylaluminium-mu-methylenetitanium

44506.AA 44506.AC
44506.AAEA 257 EUR
44506.AA 44506.AC
Tebbe Reagent 0.5 M in toluene
Tebbe Reagent

CAS No.: 67719-69-1 Molecular Formula: C13H18AlClTi Formula Weight: 284.62 Flash Point: 1°(34°F) Physical Form: Liquid Density: 0.927 Sensitivity: Air and Moisture Sensitive Appearance: Dark red-brown UN No.: UN3399 MDL No.: MFCD00151575

Kaava: C₁₃H₁₈AlClTi
MW: 284.62 g/mol
Tiheys: 0.927
Leimahduspiste: 1 °C (34 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00151575
CAS-numero: 67719-69-1
UN: 3399
ADR: 4.3 II
Merck Index: 14,9091

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR