3-Hydroksi-5-metyyli-isoksatsoli ≥97%

Toimittaja: Thermo Fisher Scientific
Vaara

44450.06 44450.14 44450.22
44450.06EA 119 EUR
44450.06 44450.14 44450.22
3-Hydroksi-5-metyyli-isoksatsoli ≥97%
3-Hydroksi-5-metyyli-isoksatsoli

Purity: 97% CAS No.: 10004-44-1 Molecular Formula: C4H5NO2 Formula Weight: 99.09 Physical Form: Powder Solubility: Soluble in alcohol, acetone, THF, chloroform MDL No.: MFCD00144468

Kaava: C₄H₅NO₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00144468
CAS-numero: 10004-44-1
EINECS: 233-000-6
Merck Index: 14,4856

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR