Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)

Toimittaja: Thermo Fisher Scientific
Varoitus

44446.02 44446.04 44446.77
44446.02EA 494 EUR
44446.02 44446.04 44446.77
Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)
Di-µ-bromobis(tri-tert-butylphosphine)dipalladium(I)

CAS No.: 185812-86-6 Molecular Formula: C24H54Br2P2Pd2 Formula Weight: 777.29 Storage Temperature: -30°C to -10°C Physical Form: Powder Sensitivity: Air Sensitive Appearance: Dark green Solubility: Soluble in benzene and toluene MDL No.: MFCD04114019


Applications: Coupling reactions. Will activate aryl chloride and sterically hindered or electron rich aryl/vinyl bromides and iodides. Especially active in difficult aminations

Kaava: C₁₂H₅₄Br₂P₂Pd₂
MDL numero: MFCD04114019
CAS-numero: 185812-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR