Pii ≥98%, crystalline powder APS ≤50 nm, laser synthesised from vapor phase

Toimittaja: Thermo Fisher Scientific
Varoitus

44384.09 44384.04 44384.18
44384.09EA 681 EUR
44384.09 44384.04 44384.18
Pii ≥98%, crystalline powder APS ≤50 nm, laser synthesised from vapor phase
Pii

Purity: 98% CAS No.: 7440-21-3 Physical Form: S.A. 70-100m2/g Appearance: White Odour: Odourless UN No.: 1346 MDL No.: MFCD00085311

Kaava: Si
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00085311
CAS-numero: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III
Merck Index: 12,08635

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR