Platina 5% in graphite Type 286

Toimittaja: Thermo Fisher Scientific
Varoitus

44365.06 44365.14 44365.22
44365.06EA 242 EUR
44365.06 44365.14 44365.22
Platina 5% in graphite Type 286
Platina

CAS No.: 7440-06-4 Physical Form: Powder Appearance: Dark grey Odour: Odourless UN No.: UN1325 MDL No.: MFCD00011179


Applications: Material for gas sensors

Kaava: Pt
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011179
CAS-numero: 7440-06-4
EINECS: 231-116-1
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR