Alumiini ≥99.9999% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)

Toimittaja: Thermo Fisher Scientific

44328.06 44328.14 44328.22
44328.06EA 146 EUR
44328.06 44328.14 44328.22
Alumiini ≥99.9999% (metals basis), slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Alumiini

Purity: 99.9999% CAS No.: 7429-90-5 Physical Form: ≈0.54g/slug Odour: Odourless MDL No.: MFCD00134029

Kaava: Al
Kiehumispiste: 2327 °C (1013 hPa)
Sulamispiste: 660 °C
Tiheys: 2,7 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00134029
CAS-numero: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR